Would you like to log in again?
Small Capacity Glas-Col® Stirmantle, Ace Glass
Supplier: Ace Glass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Glas-Col®
12046-10 12046-01 12046-27 12046-15 12046-02 12046-13 12046-17 12046-20 12046-25 12046-23
76214-474EA 1719.02 USD InStock
76214-474 76214-486 76214-484 76214-478 76214-476 76214-488 76214-492 76214-482 76214-490 76214-480
Small Capacity Glas-Col® Stirmantle, Ace Glass
Heaters Heating Mantles
Mantle stirring system with coil induced Synchostart magnetic field, exclusive to Glas-Col.
  • Variable speed StirControl II controller has a 4 foot cord and can be placed outside the hood
  • StirControl II features two receptacles for operating two Stirmantles at once
  • Complete system includes mantle, StirControl II, PTFE stirbar, and cord
Stirmantle with cords only does not include StirControl II or PTFE stirbar, for heating needs.
Ordering information: 230 V products are not returnable.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty