Would you like to log in again?
MISSION® Genomics Lentivirus Packaging Mix Powered
Supplier: Mirus Bio
MISSION® Genomics
MIR6630 MIR6640
76211-034EA 2130.49 USD InStock
76211-034 76218-672
MISSION® Genomics Lentivirus Packaging Mix Powered
Transfection Reagents
Aqueous solution for enhanced lentiviral transductions in target cell types. Provided at 10 mg/ml.
  • Produce 2 to 3x higher functional titers
  • Achieve even higher titers and eliminate the need to concentrate
  • No media change required
  • Single harvest
Accelerate viral production by skipping tedious but critical multivariable plasmid optimization. Produce high titer lentivirus with the TransIT®-Lenti Transfection Reagent. Compatible with second or third generation transfer vector encoding gene of interest (GOI). Choose between small or large scale lentivirus production with our conveniently sized 5 or 34 reaction sizes for 10cm dish. Optimized formulation of two plasmids expressing the structural and regulatory (gag, pol, and rev) and viral envelope (VSV-G) proteins required for producing high titer lentivirus with broad viral tropism.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty