Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,5-Dihydroxybenzoic acid ≥99.0% (by GC, titration analysis)

Supplier: TCI America
Synonyms: 2,5-DHBA2,5-Dihydroxybenzoic AcidGentisic acidHydroquinonecarboxylic acidDHB

D2933-1G D2933-5G
76219-014EA 37.94 USD InStock
76219-014 TCD2933-5G
2,5-Dihydroxybenzoic acid ≥99.0% (by GC, titration analysis)
2,5-Dihydroxybenzoic acid
[Matrix for MALDI-TOF/MS]
CAS Number: 490-79-9
MDL Number: MFCD00002460
Molecular Formula: C7H6O4
Molecular Weight: 154.12
Purity/Analysis Method: 99.0% (GC,T)
Form: Crystal
Melting point (°C): 206

Product Categories: Analytical Science, Standards for Mass Spectrometry & Matrix Materials
Other Product Names / Synonyms: Gentisic Acid


Formula: C₇H₆O₄
MW: 154.12 g/mol
Melting Pt: 206 °C
MDL Number: MFCD00002460
CAS Number: 490-79-9
https://us-legacy.vwr.com/stibo/web/std.lang.all/00/79/28980079.jpg
Order Now
Specification Test Results
Purity/Analysis Method 99.0% (GC,T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty