Would you like to log in again?
PAPR UNIT TR-302N+ VERSAFLO 6HRS BEIGE
Supplier: 3M OCCUPATIONAL HEALTH
Versaflo* PAPR Unit TR-302N+^ Versaflo* PAPR Unit TR-302N+, Color: Beige, Economy Battery, 6Hours battery life, Back-Mounted Breathing Tube, Head Cover with Suspension, NIOSH Approved Standards, Two selectable flow rate options 6.5CFM and 7.2CFM, Easy-to-use and versatile PAPR
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
PAPR UNIT TR-302N+ VERSAFLO 6HRS BEIGE 76220-724 7100153809 Case of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty