Would you like to log in again?
PAPR ASSEMBLY TR-304N+ VERSAFLO 6HRS CS1
Supplier: 3M OCCUPATIONAL HEALTH
Versaflo* PAPR Assembly TR-304N+^ Versaflo* PAPR Assembly TR-304N+, with Easy Clean Belt and Economy Battery, 6Hours battery life, Color: Beige, Back-Mounted Breathing Tube, Head Cover with Suspension, Two selectable flow rate options 6.5CFM & 7.2CFM offers more options and comfort to the user, CS1
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
PAPR ASSEMBLY TR-304N+ VERSAFLO 6HRS CS1 76220-728 7100153811 Case of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty