Would you like to log in again?
PAPR ASSEMBLY TR-306N+ VERSAFLO 10-12 HR
Supplier: 3M OCCUPATIONAL HEALTH
Versaflo* PAPR Assembly TR-307N+^ Versaflo* PAPR Assembly TR-307N+, with Easy Clean Belt and High Capacity Lithium Ion- Rechargeable Battery, Color: Beige, 10-11 Hours battery life, Back-Mounted Breathing Tube, Head Cover with Suspension, Two flow rate options 6.5CFM and 7.2CFM offers comfort
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
PAPR ASSEMBLY TR-306N+ VERSAFLO 10-12 HR 76220-732 7100153813 Case of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty