Would you like to log in again?
FRAME OVER VISOR VERSAFLO RADIANT HEAT
Supplier: 3M OCCUPATIONAL HEALTH
Versaflo*Over-visor Frame, M-961N^ Versaflo*Over-visor Frame, M-961, have a low profile, designed specifically for use with select helmets, allowing the user the flexibility to have tinted IR/UV protection when needed, protect for head, neck and shoulders against radiant heat, sparks and splashes.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
FRAME OVER VISOR VERSAFLO RADIANT HEAT 76220-746 7100154127 Case of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty