Would you like to log in again?
BATTERY TR-830/94243(AAD) VERSAFLO SAFE
Supplier: 3M OCCUPATIONAL HEALTH
Versaflo* Battery TR-830/94243(AAD) for TR-800 PAPR^ Versaflo* Battery TR-830/94243(AAD), Intrinsically Safe, for TR-800 PAPR, IS protection - rated at Division 1 for Classes I,II,and III under the most current UL standard, Combine with certain other Versaflo* components to build a complete system
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
BATTERY TR-830/94243(AAD) VERSAFLO SAFE 76220-750 7100150925 Case of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty