Would you like to log in again?
RESPIRATOR UNIT PURIFYING VERSAFLO CS1
Supplier: 3M OCCUPATIONAL HEALTH
Versaflo* Respirator Unit TR-802N/94242(AAD)^ Versaflo*Powered Air Purifying Respirator Unit TR-802N/94242(AAD), Intrinsically Safe, IS protection - rated at Division 1 for Classes I,II,and III under the current UL standard, Combine with other Versaflo* components to build a complete system, CS 1
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
RESPIRATOR UNIT PURIFYING VERSAFLO CS1 76220-768 7100150821 Case of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty