Would you like to log in again?
MastAssure™ Bacterial Agglutinating Antisera, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
M11002 M10326 M10342 M11303 M10345 M10330 M10113 M10344 M10319 M10150 M10346 M10340 M10309 M10111 M10109 M10365 M10335 M10336 M10154 M10347 M10312 M10343 M92537 M10376 M10134 M10114 M10112 M10133 M10367 M10341 M10313 M11302 M12030 M10320 M14300HD M10132 M10127 M10331 M10311 M10314 M10310 M10702 M10332 M91875 M10308 M10318 M10364 M10110 M10115 M10377 M10157 M14294 M11702 M10126 M10128
76220-850EA 108.54 USD InStock
76220-850 76220-894 76220-892 76220-890 76220-858 76220-812 76220-856 76220-810 76220-854 76220-898 76220-852 76220-896 76220-806 76220-848 76220-804 76300-040 76220-882 76220-880 76220-846 76220-802 76220-844 76220-888 76220-800 76220-886 76220-842 76220-840 76220-884 76220-918 76220-916 76220-838 76220-872 76220-870 76220-912 76220-836 76220-910 76220-834 76220-798 76220-832 76220-830 76220-908 76220-828 76220-906 76220-826 76220-904 76220-860 76220-824 76220-868 76220-902 76220-822 76220-866 76220-900 76220-820 76220-864 76220-862 76220-816
MastAssure™ Bacterial Agglutinating Antisera, Hardy Diagnostics
Media for Microbiology Additives and Supplements for Microbiology
An extensive range containing both polyvalent and monovalent diagnostic antisera for the identification of clinical bacteria according to the expression of somatic antigens.
  • High quality performance
  • Extensive range available
  • Easy to read
  • Reliable identification
Bacterial serotype can be determined by both slide agglutination and test tube agglutination methods.
The antisera are derived from immunised rabbits, adsorbed to remove cross agglutinins and filter sterilized.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty