Would you like to log in again?
Oakton® Ion 700 Benchtop pH and ISE Meter, Cole-Parmer
Supplier: Cole Parmer
35419-23 WD-35419-20 WD-35419-22
76221-220EA 1000.94 USD InStock
76221-220 101076-310 76221-218
Oakton® Ion 700 Benchtop pH and ISE Meter, Cole-Parmer
Electrochemical Meters pH and Multi-Parameter Meters
Designed for today's crowded laboratory benchtops.
  • Takes up less bench space—40% smaller than comparable models
  • Super-large dual-line display is easy to read
  • Simple calibrations with 5-point automatic pH buffer recognition for USA and NIST buffers
  • More accurate measurements with up to 5-point calibration for ion concentration on Ion 700 model
  • Versatile—use with any pH, ORP, or ISE electrodes with BNC connector
Ion 700 meter measures pH, mV, relative mV, and temperature, like a standard benchtop pH meter. When used with ISE's for ion concentration measurements, it also reads directly in ppm for easy use. Automatic temperature compensation (ATC) maintains reading accuracy even with fluctuating temperatures.
Meters feature Auto-Hold, selectable °F or °C measurement, and easy recall of electrode slope or offset.
Certifications: CE Approved; 3 Year Meter Warranty; ISO9001:2008.
Ordering information: Model WD-35419-20 includes meter with universal AC power adapter, one double-junction pH electrode, one temperature (ATC) probe, and a removeable electrode holder. For replacement parts, see model WD-35613-13 for the Automatic Temperature Compoensation (ATC) probe, model WD-35641-51 for a pH electrode, model WD-35615-07 for the universal power adapter, and model WD-35420-54 for the electrode holder.
Model WD-35419-22 includes meter with universal AC power adapter, and a removeable electrode stand. For replacement parts, see model WD-35615-07 for the univerrsal power adapter, and model WD-35420-54 for the electrode holder.
Model WD-35419-23 includes meter with universal AC power adapter, and one All-in-One pH/ATC electrode. For replacement parts, see model WD-35811-71 for the probe, model WD-35615-07 for the univerrsal power adapter, and model WD-35420-54 for the electrode holder.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty