Would you like to log in again?
Oakton® pH 2700 Benchtop Meters, Cole-Parmer
Supplier: Cole Parmer
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Oakton®
35420-20 35420-22
76221-224EA 2092.2 USD InStock
76221-224 76221-226
Oakton® pH 2700 Benchtop Meters, Cole-Parmer
Electrochemical Meters pH and Multi-Parameter Meters
Microprocessor based pH 2700 benchtop meter measures pH, mV, relative mV, and temperature.
  • Large display with bright backlight
  • High 0.001 pH resolution with ±0.002 pH accuracy
  • Password protection for both setup and calibration for security
  • Calibration-due alarm let you know when to recalibrate
  • Audible alarms alert you when reading fall out of range
  • Store and recall up to 500 sets of data, send data to a PC or printer
  • Optional password protection prevents changes to meter setup and calibration
Meters automatically calculate and display electrode slope and offset, ideal for checking electrode quality.
Nonvolatile memory stores up to 500 data sets to simplify documenting large volumes of pH or mV measurements. Meters automatically calculate and display electrode slope and offset - ideal for checking electrode quality. Advanced set-up options allow for a large degree of user customization.
Advanced set-up options allow for a large degree of user customization. Datalogging intervals can be programmed into the meter for convenient continuous monitoring. Programmable limit alarm alerts you when readings fall out of range. Calibrate up to 6 points for pH using USA, NIST, DIN, or custom buffer sets.
Informative display shows pH or redox measurements together with temperature, electrode status, cal points, time and date all at once. Cal-due alarm lets you know when recalibration is necessary for measurement accuracy. Visual stability indicator depicts measurements have stabilized thereby eliminating guesswork.
Certifications: It meets GLP requirements - meters stamp all calibration and stored data with time/date.
Ordering information: Order RS232 and optional RS232 to USB adapter separately.
Delivery information: Includes electrode stand and 100/240 VAC power adapter.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty