Would you like to log in again?
Oakton® Waterproof PC 450 pH/CON Portable Meters Antylia Scientific
Supplier: Antylia Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Oakton®
35630-90 35630-10 35630-12 35630-80 35630-32
76221-286EA 1960.77 USD InStock
76221-286 76221-288 76221-282 76221-280 76221-278
Oakton® Waterproof PC 450 pH/CON Portable Meters Antylia Scientific
Electrochemical Meters pH and Multi-Parameter Meters
More measurements from one meter.
  • Get both pH and conductivity measurements with combination pH/CON probe or separate pH and conductivity probes
  • Use it in any environment with backlit display and rugged design
  • Intuitive, user-friendly icons show electrode status as pH slope and all constant
  • Get quick and easy electrode attachment to a beaker or container with Grip-Clip™ holder
  • Download and analyze up to 500 data sets with USB or RS232 output
  • Use as a benchtop meter as well—built-in stand and wall-mount option
Ideal meter for any environment with its rugged, waterproof housing. View both the main measurement and temperature simultaneously with large, backlit LCD. Ensure the accuracyof your measurements with up to six calibration points from USA, NIST, DIN, and custom pH buffer sets. Autocal feature for conductivity has one calibration point per range. Don’t worry about accuracy in fluctuating temperature, ATC ensures measurement accuracy.
Meter features include ready (stability) indicator, smart averaging, user-selectable °C/°F units, hold function, calibration alarm, battery-life indicator, and electrode status. Choose combination pH/CON probe or separate probe options.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty