Would you like to log in again?
Tyramide Amplification Kit with HRP Goat Anti-Rabbit and CF® Dye or Biotin Tyramide, Biotium
Supplier: Biotium
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
33019 33010 33001 33004 33007 33016 33013
76221-398EA 723.74 USD InStock
76221-398 76221-404 76221-416 76221-428 76221-410 76221-422 76221-798
Tyramide Amplification Kit with HRP Goat Anti-Rabbit and CF® Dye or Biotin Tyramide, Biotium
Protein Linking and Labeling Systems
Kits for increasing immunofluorescence sensitivity using HRP-catalyzed tyramide signal amplification with the choice of next-generation fluorescent CF® dyes or biotin.
  • Increase sensitivity by up to 100-fold
  • Use lower primary antibody concentrations
  • Boost signal in challenging samples, like human FFPE tissues
  • Choose from six bright and photostable CF® dyes or biotin
Tyramide amplification, sometimes called Catalyzed Reporter Deposition (CARD), is an enzyme-mediated detection method that uses the catalytic activity of horseradish peroxidase (HRP) to generate high density labeling of a target protein or nucleic acid sequence in situ. The tyramide amplification method has been reported to increase the sensitivity by up to 100-fold compared to conventional avidin-biotin detection. Multiple rounds of tyramide signal amplification can be performed for multiplex detection.
The HRP goat anti-rabbit antibody provided in the kits has been adsorbed against human, mouse, and rat serum to minimize cross-reactivity. The kits include choice of six CF® dye tyramides or biotin tyramide. CF® dyes are line of next-generation fluorescent dyes, with advantages in brightness and photostability compared to other dyes.
Ordering information: Each kit contains reagents for staining 50 to 150 slides; the actual number of samples that can be stained will depend on staining volumes used. Tyramide amplification buffer, CF® dye tyramides, and other tyramide compounds are also available separately. Note: Endogenous biotin blocking reagents (not provided) may be required for streptavidin/biotin detection in cells or tissues.
Delivery information: Kit components: CF® dye or biotin-tyramide stock solution, HRP goat anti-rabbit IgG (highly cross-adsorbed), BSA for blocking, Tyramide amplification buffer plus, 30% Hydrogen peroxide.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty