Would you like to log in again?
dUTP CF® Dye Conjugates, Biotium
Supplier: Biotium
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
40007-T 40100-T 40004 40002 40005 40006 40100 40008-T 40005-T 40003-T 40003 40008 40004-T 40006-T 40007 40002-T
89410-682EA 432.08 USD InStock
89410-682 76221-628 89410-680 89410-678 89410-676 71004-056 89410-674 89411-562 76221-442 76221-634 76221-744 76221-624 76221-636 76221-626 76221-630 76221-632
dUTP CF® Dye Conjugates, Biotium
Protein Linking and Labeling Systems
CF® dye-dUTP can be used for TUNEL assay, microarray, or to synthesize labeled DNA probes for in situ hybridization and nucleic acid blotting applications.
  • For labeling fluorescent DNA probes
  • Detect by fluorescence microscopy, flow cytometry, or in gel
  • Compatible with fixed cells or tissue sections
  • Choice of 8 CF® dye colors from blue to near infrared
  • Biotium’s CF® dyes have superior brightness, photostability, and are water-soluble
Biotium’s next-generation CF® dyes were designed to be highly water-soluble with advantages in brightness and photostability compared to other fluorescent dyes. CF® dye dUTP conjugates are supplied in lyophilized form.
Caution: For PCR applications, Taq polymerase should be used with dUTP conjugates, because dUTP inhibits archaeal polymerases such as Pfu and Vent®.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty