Would you like to log in again?
ViaFluor® 405 Live Cell Microtubule Stain 1000X in DMSO, Biotium
Supplier: Biotium
70064 70064-T
76221-658EA 126.88 USD InStock
76221-658 76221-446
ViaFluor® 405 Live Cell Microtubule Stain 1000X in DMSO, Biotium
Assays Cellular Assays
Stain microtubules in live cells with blue fluorescence.
  • Cell permeable fluorescent taxol probes
  • Rapid, sensitive, no-wash staining
  • Blue fluorescence for the DAPI channel
  • Ex/Em: 408/452 nm
  • Available with blue, green, or far-red fluorescence
ViaFluor® Live Cell Microtubule Stains are suitable for imaging the microtubule cytoskeleton in live cells.
Taxol binds to polymerized tubulin and stabilizes microtubules, resulting in inhibition of mitosis. However, fluorescent taxol compounds like ViaFluor® Microtubule Stains are less disruptive of microtubule dynamics and cell division, presumably due to lower binding affinity of the fluorescent probe compared to taxol itself. The probes can be used to stain cells for as long as 48 hours to several days. However, the probes may show cytotoxicity and cell cycle arrest with longer incubation times. ViaFluor® Live Cell Microtubule Stains cannot be fixed after staining and cannot be used with fixed cells or tissues. The stains are supplied as a 1000X stock solutions in DMSO. They come with a vial of 100 mM verapamil, an efflux pump inhibitor that may improve probe retention and staining in some cell types.
Briefly centrifuge vials to collect contents at the bottom of the tubes before opening.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty