Would you like to log in again?
Lysosomal Stain, LysoView™, Biotium
Supplier: Biotium
LysoView™
70058 70061-T 70066-T 70061 70067 70066 70067-T
76221-450EA 220.17 USD InStock
76221-450 76221-452 76221-662 75832-696 10066-564 75832-698 76221-664
Lysosomal Stain, LysoView™, Biotium
Assays Cellular Assays
LysoView™ dyes label lysosomes in live cells, and are available with blue, green, visible red, and far-red fluorescence.
  • No-wash, fluorescent staining of lysosomes in live cells
  • Non-toxic for real-time live-cell imaging
  • Lower background than LyosoTracker® dyes
  • Dye options for super-resolution imaging
  • Blue, green, red, or far-red fluorescence
LysoView™ dyes are fluorescent stains for imaging lysosome localization and morphology in live cells. The dyes accumulate in the low pH environment of the lysosomes, resulting in highly specific lysosomal staining without the need for a wash step. LysoView™ 540 and LysoView™ 633 exhibit pH-sensitive fluorescence, further enhancing the specificity of staining. LysoView™ 488 has been validated in super-resolution imaging by SIM, while LysoView™ 650 fluorophore is compatible with super-resolution imaging by SIM and STED.
LysoView™ 540 and LysoView™ 633 stain vacuoles in yeast. Staining of bacteria varies between dyes.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty