Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

SYBR® Green I 10000x in DMSO nucleic acid gel stain

Supplier: Biotium

Warning
Synonyms: Thiazole Green

40086-0.5ML 40086-1ML
76221-740EA 203.32 USD InStock
76221-740 76221-742
SYBR® Green I 10000x in DMSO nucleic acid gel stain
SYBR® Green I
Thiazole green, which is structurally identical to SYBR® green I nucleic acid gel stain, is one of the most sensitive stains available for detecting double-stranded DNA (dsDNA) in agarose, polyacrylamide gels and qPCR.

  • Color & Form Orange solution
  • Ex/Em 497/520nm (with DNA)

Thiazole green can also be used to detect ssDNA and RNA in denaturing agarose/formaldehyde and polyacrylamide/urea gels without any prewashing steps, although the sensitivity is lower. Thiazole green has the identical spectral properties as SYBR® green I (Ex/Em 497/520 nm with DNA).

SYBR is a registered trademark of Thermo Fisher Scientific.


Formula: C₃₂H₃₇N₄S
MW: 509.73 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00284715
CAS Number: 163795-75-3
Order Now


 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty