Would you like to log in again?
VWR CYLINDER MEASURING CL A TPX 25ML BX2
Supplier: VWR International
VWR* Measuring Cylinder, Class A, Material: TPX, Autoclavable, volume: 25 ml, Box of 2^
ORDER
Expand All / Collapse All
Description VWR Catalog Number Unit Price Quantity
VWR CYLINDER MEASURING CL A TPX 25ML BX2 76221-758 Box of 2 Retrieving
Case of 16 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
DOCUMENTATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty