Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

BTX ECM 2001+ Electrofusion and Electroporation Generators, Harvard Apparatus

Supplier: HARVARD APPARATUS
BTX
45-2045 45-2049 45-2048 45-2050 45-2047 45-2046
76221-826EA 37569.64 USD InStock
76221-826 76221-828 76221-822 76221-824 76221-830 76221-820
BTX ECM 2001+ Electrofusion and Electroporation Generators, Harvard Apparatus
Electroporation Systems Electroporators
ECM 2001+ instruments offer best-in-class features for cell fusion and transfection applications.

  • AC waveform of 0.2 - 2.0 MHz
  • Square wave mammalian electroporation capabilities
  • Wide range of voltages from 5 V to 3000 V
  • Pre-set protocols include most common cell fusion applications
  • User-defined protocols allow unlimited ability to add and modify protocols
  • Advanced programmability, combines up to 19 AC steps pre- and post- DC fusion step
  • Simple to use, large 7" touchscreen interface

The ECM 2001+ is a multifunctional electrofusion and square wave electroporation generator. The ability to generate both AC and DC waves allows for fast and efficient cell fusion in hybridoma production, hybrid cell formation, and nuclear transfer applications. This system is powerful enough to yield high transfection efficiencies for cell lines and difficult to transfect cell types including stem cells and primary cells. The gentle square wave pulse also allows for high cell viability of these cell types.

Fast, efficient cell fusion in hybridoma production, hybrid cell formation, and nuclear transfer applications is facilitated by the combination of AC and DC wave pulses. Fusion is achieved by the generation of an AC current wave form that generates a benign dielectrophoretic alignment of cells. With a 30 ms switchover time from AC to DC, efficient fusion takes place. After fusion, the AC is reapplied maintaining the cell compression for the rounding off process, resulting in a higher number of hybrids.

For use in large-scale hybridoma production applications, the ECM 2001+ generator, 2 ml coaxial optimization or 9 ml coaxial production chambers, high voltage output cable, and female/female adapters may be used. Alternatively, for medium-scale hybridoma production applications, the ECM 2001+ generator may be combined with 650 ml, 3.2 mm gap or 2 ml, 10 mm gap microslides, high voltage output cable, and micrograbber adapters. This system does not require proprietary fusion medium, however may be used with the Cytofusion Medium C for enhanced efficiency, reproducibility, and convenience.

Certifications: CE, ETL, WEEE marked (UL, CSA). IECEE CB Scheme certified. EU RoHS compliant.

Accessories information: BTX cables and adapters are needed to connect an electrofusion/electroporation system to the electordes and chambers used for a particular application. A single High Voltage cable is used specifically for connecting the Legacy ECM 2001 to accessories. The High Voltage output cable is available in either 1.5 or 3 meters in length. The cable has a coaxial attachment with interlock grounding cable to the instrument and red and black banana attachments to the desired accessory.

Order Now
 

ORDER

ACCESSORIES

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty