Would you like to log in again?
Llama IgG1 isotype control (Biotin)
Supplier: Rockland Immunochemicals
025-0640
76221-906EA 195 USD InStock
76221-906
Llama IgG1 isotype control (Biotin)
Antibodies
Llama IgG1 isotype control Biotin Conjugate
  • control or standard reagent
  • ELISA, western blotting, Immunohistochemistry
Comparative studies of old world camelids (Camelus bactrianus and Camelus dromedarius) and new world camelids (Lama pacos, Lama glama and Lama vicugna) have shown that heavy-chain-only immunoglobulins represent between 35% - 70% of total IgG in the sera of all species. Such antibodies are homodimers of heavy chains that lack the CH1 domain of conventional antibodies and therefore do not interact with light chains, exhibiting a lower molecular weight ~100 kDa. In llama and other species of camelids, these heavy-chain-only immunoglobulins belong to the IgG2 and IgG3 subclasses.
All gamma chain camelid antibodies exhibiting the more conventional assembly of two light and two heavy chains with molecular weight ~150 kDa, belong to the IgG1 subclass.
Llama IgG1 Biotin Conjugated isotype control can be utilized as a control or standard reagent in Flow Cytometry, ELISA, and western blotting experiments where determination of sample isotype is important.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty