Would you like to log in again?
Llama IgG3 isotype control (FITC (Fluorescein Isothiocyanate))
Supplier: Rockland Immunochemicals
025-0243
76221-916EA 195 USD InStock
76221-916
Llama IgG3 isotype control (FITC (Fluorescein Isothiocyanate))
Antibodies
Llama IgG3 isotype control Fluorescein Conjugate
  • control or standard reagent
  • multiplex, immunofluorescence microscopy, fluorescence based plate assays (FLISA) and fluorescent western blotting
Comparative studies of old world camelids (Camelus bactrianus and Camelus dromedarius) and new world camelids (Lama pacos, Lama glama and Lama vicugna) have shown that heavy-chain-only immunoglobulins represent between 35% - 70% of total IgG in the sera of all species. Such antibodies are homodimers of heavy chains that lack the CH1 domain of conventional antibodies and therefore do not interact with light chains, exhibiting a lower molecular weight ~100 kDa.
In llama and other species of camelids, these heavy-chain-only immunoglobulins belong to the IgG2 and IgG3 subclasses of gamma chain antibodies.
Llama IgG3 FITC conjugated isotype control can be utilized as a control or standard reagent in Flow Cytometry, immunofluorescence microscopy, fluorescence based plate assays (FLISA) and fluorescent western blotting experiments where determination of sample isotype is important.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty