Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

GENIE 100™ Temp-Shaker, 230 V

Supplier: Scientific Industries
SI-G102 SI-G103 SI-G104 SI-G101
76226-004EA 4829.73 USD InStock
76226-004 76226-092 76226-008 76226-006
GENIE 100™ Temp-Shaker, 230 V
Shakers and Mixers Shaking Incubators
Gentle Orbital Shaking from 20 to 100 rpm, while providing precise temperature control from 28 to 75 °C, for use in many laboratory applications.

  • 230 V model
  • Unique three-way access clear dome chamber provides 360° view at all times
  • Quick access to your samples with easy tilt-n-rest feature using the exclusive 'kick stand' feature to open the chamber cover, left, right, or back, to optimize bench space
  • The lightweight cover can also be easily removed for cleaning and attaching accessories
  • Unique airflow design can heat the chamber to a stable 75 °C in as little as 20 minutes
  • With the chamber door open, unit will maintain the chamber temperature within ±3 °C

Various voltages and plugs available for operation around the world. Product is produced and shipped from within the US.

Accessories information: No additional accessories necessary for operation of this model. However, stacking tray is available to double capacity. No tools necessary to mount accessories.

Delivery information: This model includes the non-slip tray with drip-lip to contain any spills. Non-slip stacking tray and additional interchangeable platform styles available, sold as accessories.

Caution: 230 V model. Corresponding model with voltage suitable for use in US (76226-090) is also available, please see separate product page.

Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty