Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

GENIE 300™ Temp-Shaker, 230 V

Supplier: Scientific Industries
SI-G1501 SI-G1704 SI-G1703 SI-G1502 SI-G1601 SI-G1604 SI-G1701 SI-G1602 SI-G1504 SI-G1503 SI-G1702 SI-G1603
76226-034EA 5634.63 USD InStock
76226-034 76226-044 76226-054 76226-032 76226-042 76226-038 76226-048 76226-028 76226-052 76226-030 76226-040 76226-050
GENIE 300™ Temp-Shaker, 230 V
Shakers and Mixers Shaking Incubators
High speed orbital shaking from 35 to 300 rpm, while providing precise temperature control from 28 to 75 °C, for use in many laboratory applications.

  • 230 V model
  • Unique 3-way access clear dome chamber provides 360° view at all times
  • Quick access to your samples with easy tilt-n-rest feature using the exclusive 'kick stand' feature to open the chamber cover, left, right, or back, to optimize bench space
  • The lightweight cover can also be easily removed for cleaning and attaching accessories
  • Unique airflow design can heat the chamber to a stable 75 °C in as little as 20 minutes
  • With the chamber door open, unit will maintain the chamber temperature within ±3 °C

Various voltages and plugs available for operation around the world. Product is produced and shipped from within the US.

Accessories information: Flask Clamps model requires flask clamps sold separately. All platforms are easily interchangeable.

Delivery information: The Genie Temp-Shaker 300 series models include the required platform.

Caution: 230 V model. Corresponding model with voltage suitable for use in US (76226-010, 76226-036 and 76226-046) is also available, please see separate product page.

Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty