Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgG Rabbit Polyclonal Antibody (DL800)

Supplier: Rockland Immunochemicals
617-445-002
76226-082EA 130 USD InStock
76226-082
Anti-IgG Rabbit Polyclonal Antibody (DL800)
Antibodies
Conjugated Anti-Monkey IgG (H&L) DyLight™ 800 Conjugated antibody generated in rabbit detects specifically monkey IgG heavy and light chains. Secreted as part of the adaptive immune response by plasma B cells, immunoglobulin G constitutes 75% of serum immunoglobulins. IgG binds to viruses, bacteria, as well as fungi and facilitates their destruction or neutralization via agglutination (and thereby immobilizing them), activation of the compliment cascade, and opsinization for phagocytosis. The whole IgG molecule possesses both the F(c) region, recognized by high-affinity Fc receptor proteins, as well as the F(ab) region possessing the epitope-recognition site. Both heavy and light chains of the antibody molecule are present. This DyLight™800 conjugated anti-Monkey IgG (H&L) secondary antibody is ideal for investigators who routinely perform immunofluorescence, flow cytometry, and more general immunoassays. When choosing a secondary antibody product, consideration must be given to species and immunoglobulin specificity, conjugate type, fragment and chain specificity, level of cross-reactivity, and host-species source and fragment.The emission spectra for this DyLight™ conjugate match the principle output wavelengths of most common fluorescence instrumentation.

Anti-Monkey IgG (H&L) DyLight™ 800 conjugated Antibody is suitable for use in immunofluorescence microscopy, fluorescence based plate assays (FLISA) and fluorescent western blotting. This product is also suitable for multiplex analysis, including multicolor imaging, utilizing various commercial platforms. Specific conditions for reactivity and signal detection should be optimized by the end user.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DL800
Epitope:
Host: Rabbit
Isotype:
Reactivity: Monkey

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty