Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgG1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
634-401-044
76226-086EA 220 USD InStock
76226-086
Anti-IgG1 Rabbit Polyclonal Antibody
Antibodies
Camelids produce functional IgG isotypes that do not incorporate light chains. Comparative studies of old world camelids (Camelus bactrianus and Camelus dromedarius) and new world camelids (Lama pacos, Lama glama and Lama vicugna) have shown that heavy-chain-only immunoglobulins represent between 35% - 70% of total IgG in the sera of all species. At present, three subclasses of camelid IgG have been identified (IgG1, IgG2, IgG3), of which IgG2 and IgG3 lack the light chains. IgG2 consists of dimers of short heavy chains, which are characterized by a normal Fc region without a CH1 domain. IgG2 shows a molecular weight of approximately 90 kDa and this smaller size improves its bio-distribution and allows a better tissue penetration. Of special importance is the long complementary determining region (CDR) loop which inserts deep into the active site of an enzyme enabling a complete neutralization of an enzyme. Anti-Llama IgG2 generated in rabbit detects specifically Llama IgG2 isotype. This anti-Llama secondary Antibody is suitable for western blot, ELISA, ChIP and immunohistochemistry as well as other more general immunoassays.

Antibody Anti-Llama IgG2 is suitable for immunoblotting (western or dot blot), ELISA, and immunohistochemistry assays requiring lot-to-lot consistency.

Type: Secondary
Antigen: IgG1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Camel

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty