Would you like to log in again?
RAT SERUM (sterile) (Heat Inactivated), Rockland Immunochemicals
Supplier: Rockland Immunochemicals
D110-10-0500 D110-10-0050 D110-10-1000 D110-10-0100
76226-484EA 6567.08 USD InStock
76226-484 76226-482 75935-198 76226-480
RAT SERUM (sterile) (Heat Inactivated), Rockland Immunochemicals
Sera (Serum)
Heat Inactivated Sterile Serum
  • 0.22 micron filter and found to be negative for bacterial growth after incubation on trypticase soy agar for 24 hours, 48 hours, and 72 hours
  • prepared by removal of fibrinogen through clot formation
  • separated from cellular components and purified through centrifugation and filtration techniques
Rat Serum is used as a supplement to cell culture media. Rat Serum is also suitable for use as a component of bioassays, immunoassays or enzyme assays. Rat Serum provides a broad spectrum of macromolecules, carrier proteins for lipoid substances and trace elements, attachment and spreading factors, low molecular weight nutrients, and hormones and growth factors that promote cell growth and health. Be certain to maintain Good Cell Culture Practice, and maintain sterility of cultures that require media supplementation. Rat Serum is ideal for investigators in Cancer, Immunology, and Cell Biology research.
pH: normal
Immunoelectrophoresis: normal
Hemoglobin: normal
IgG Concentration: normal
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty