Would you like to log in again?
GOAT SERUM (sterile) (Heat Inactivated), Rockland Immunochemicals
Supplier: Rockland Immunochemicals
D104-10-0500 D104-10-1000 D104-10-0100 D104-10-0050
76230-084EA 524 USD InStock
76230-084 76230-088 76230-086 75934-356
GOAT SERUM (sterile) (Heat Inactivated), Rockland Immunochemicals
Sera (Serum)
Heat Inactivated Sterile Serum
  • 0.22 micron filter and found to be negative for bacterial growth after incubation on trypticase soy agar for 24 hours, 48 hours, and 72 hours
  • prepared by removal of fibrinogen through clot formation
  • separated from cellular components and purified through centrifugation and filtration techniques
Goat Serum is used as a supplement to cell culture media. Goat Serum provides a broad spectrum of macromolecules, carrier proteins for lipoid substances and trace elements, attachment and spreading factors, low molecular weight nutrients, and hormones and growth factors that promote cell growth and health. Be certain to maintain Good Cell Culture Practice, and maintain sterility of cultures that require media supplementation.
pH: normal
Immunoelectrophoresis: normal
Hemoglobin: normal
IgG Concentration: normal
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty