Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PECAM1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
100-401-GN4S
76234-790EA 145 USD InStock
76234-790
Anti-PECAM1 Rabbit Polyclonal Antibody
Antibodies
Anti-CD31 antibody was designed, produced, and validated as part of the Joy Cappel Young Investigator Award (JCYIA). CD31 (PECAM-1) is a 130 kDa platelet endothelial cell adhesion molecule encoded by the PECAM1 gene found on chromosome 17 in humans. CD31 is expressed on platelets, monocytes, neutrophils, and some types of T-cells, and makes up a large portion of endothelial cell intercellular junctions. CD31 plays a key role in modulation of integrin-mediated cell adhesion, leukocyte transendothelial migration, angiogenesis, apoptosis and macrophage phagocytosis. CD31 is also expressed in certain tumors, including epithelioid hemangioendothelioma, epithelioid sarcoma-like hemangioendothelioma, other vascular tumors, histiocytic malignancies, and plasmacytomas. Anti-CD31 is ideal for researchers interested in Stem Cell Research, Epigenetics, and Cell-cycle Regulation research.

CD31 antibody has been tested for use in ELISA, immunohistochemistry, and by western blot. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 80 kDa in size corresponding to CD31 protein by western blotting in the appropriate stimulated tissue or cell lysate or extract.

Type: Primary
Antigen: PECAM1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty