Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ORAI1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
100-401-P17S
76234-794EA 148 USD InStock
76234-794
Anti-ORAI1 Rabbit Polyclonal Antibody
Antibodies
The ORAI-1 gene codes for the calcium release-activated calcium (CRAC) channel subunit in humans. The CRAC channel subunit mediates calcium influx when activated by the calcium sensor, STIM1, following depletion of intracellular calcium stores. CRAC channels are the primary pathway for calcium influx in T-cells and promote the immune response to pathogens through activation of the transcription factor NFAT. Defects in this gene can be a cause of immune dysfunction due to inactivation of T-cells.

Anti-ORAI-1 has been tested for use in western blotting. Specific conditions for reactivity should be optimized by the end user. Expect bands approximately 32.7 kDa by western blotting in the appropriate cell lysate or extract.

Type: Primary
Antigen: ORAI1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty