Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IL1R1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
109-401-304-0100
76234-796EA 518 USD InStock
76234-796
Anti-IL1R1 Rabbit Polyclonal Antibody
Antibodies
IL-1 Receptor Type I is a cytokine receptor which binds interleukin 1. Anti-IL-1 Receptor Type I antibody is ideal for researchers interested in Immunology, Signal Transduction and Cancer.

This antiserum against anti-Human IL-1 Receptor Type I (IL-1RI) has been tested for use in ELISA,  radioimmunoassays and immunoprecipitation.  Reactivity in other immunoassays is unknown.This product has been assayed for the ability to immunoprecipitate antigen.  A dilution range of 1:400 to 1:800 is suggested for this immunoassay.  For immunoprecipitation, pre-clearing with a non-specific rabbit IgG is helpful to reduce background.  This product has been assayed by ELISA against soluble (extracellular) IL-1RI using HRP Conjugated Anti-Rabbit IgG [H&L] (Goat) (code # 611-1302) and ABTS as a substrate for 30 minutes at room temperature.  A working dilution range of 1:1,000 to 1:2000 is suggested for this product.  This product has been assayed by radioimmunoassay against antigen.  A dilution of 1:8,000 is suggested for this immunoassay. 

Type: Primary
Antigen: IL1R1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty