Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IL1R2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
109-401-305S
76234-802EA 148 USD InStock
76234-802
Anti-IL1R2 Rabbit Polyclonal Antibody
Antibodies
Interleukin-1 Receptor Type 2 (IL-1RII) ins a non-signaling receptor for IL1A, IL1B and IL1RN and reduces IL1B activities. It serves as a decoy receptor by competitive binding to IL1B and preventing its binding to IL1R1. It also modulates cellular response through non-signaling association with IL1RAP after binding to IL1B. IL1R2 (membrane and secreted forms) preferentially binds IL1B and poorly IL1A and IL1RN. The secreted IL1R2 recruits secreted IL1RAP with high affinity; this complex formation may be the dominant mechanism for neutralization of IL1B by secreted/soluble receptors.

This antiserum against Human IL-1 Receptor Type II (IL-1RII) has been tested for use in ELISA, radioimmunoassays and immunoprecipitation.  Reactivity in other immunoassays is unknown.  IL-1RII has been termed p68 or CD121 in some laboratories.  IL-1RII has been shown to bind IL-1, IL-1a and IL-1b.

Type: Primary
Antigen: IL1R2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty