Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TNFR-I Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
109-401-308S
76234-810EA 148 USD InStock
76234-810
Anti-TNFR-I Rabbit Polyclonal Antibody
Antibodies
Tumor necrosis factor receptor superfamily member 1A (TNF p55 Receptor) is a receptor for TNFSF2/TNF-alpha and homotrimeric TNFSF1/lymphotoxin-alpha. The adapter molecule FADD recruits caspase-8 to the activated receptor. The resulting death-inducing signaling complex (DISC) performs caspase-8 proteolytic activation which initiates the subsequent cascade of caspases (aspartate-specific cysteine proteases) mediating apoptosis. It contributes to the induction of non-cytocidal TNF effects including anti-viral state and activation of the acid sphingomyelinase.

This antiserum against Human TNF p55 Receptor (sTNFRp55) has been tested for use in ELISA,  radioimmunoassays, immunoblotting and immunoprecipitation.  Reactivity in other immunoassays is unknown.This product has been assayed for the ability to immunoprecipitate antigen.  A dilution range of 1:400 to 1:800 is suggested for this immunoassay.  For immunoprecipitation, pre-clearing with a non-specific rabbit IgG is helpful to reduce background.  This product has been assayed by immunoblot  using HRP Goat-anti-Rabbit IgG [H&L] (code # 611-1302) and TMB as a substrate.  A working dilution range of 1:200 to 1:400 is suggested for this application.  This product has been assayed by ELISA against soluble (extracellular) sTNFRp55 using HRP Conjugated Anti-Rabbit IgG [H&L] (Goat) (code # 611-1302) and ABTS as a substrate for 30 minutes at room temperature.  A working dilution range of 1:1,000 to 1:2,000 is suggested for this product.  For use in ELISA formats, this antibody is best used as the second antibody  in combination with a monoclonal antibody as a capture antibody.  This product has been assayed by radioimmunoassay against antigen.  A dilution of 1:8,000 is suggested for this immunoassay.

Type: Primary
Antigen: TNFR-I
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty