Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CCL3 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
109-401-315S
76234-820EA 148 USD InStock
76234-820
Anti-CCL3 Rabbit Polyclonal Antibody
Antibodies
C-C motif chemokine 3, CCL3 or (MIP-1a) is a monokine with inflammatory and chemokinetic properties. CCL3 binds to CCR1, CCR4 and CCR5. It is one of the major HIV-suppressive factors produced by CD8+ T-cells. Recombinant MIP-1-alpha induces a dose-dependent inhibition of different strains of HIV-1, HIV-2, and simian immunodeficiency virus (SIV).

This antiserum against Human MIP-1a has been tested for use in ELISA of human MIP-1a found in cell supernatants or in plasma or serum.  Reactivity in other immunoassays is unknown.

Type: Primary
Antigen: CCL3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty