Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Leptin Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
110-401-320S
76234-824EA 148 USD InStock
76234-824
Anti-Leptin Rabbit Polyclonal Antibody
Antibodies
Leptin is a key player in the regulation of energy balance and body weight control. Once released into the circulation, it has central and peripheral effects by binding LEPR, found in many tissues, which results in the activation of several major signaling pathways. It acts as an appetite-regulating factor, regulates bone mass and secretion of hypothalamo-pituitary-adrenal hormones, it increases basal metabolism, influences reproductive function, regulates pancreatic beta-cell function and insulin secretion, it induces FOS and SOCS3 expression to release anorexigenic peptides. It has a modulatory role in nutrient absorption. It reduces glucose absorption by enterocytes by activating PKC and leading to a sequential activation of p38, PI3K and ERK signaling pathways which exerts an inhibitory effect on glucose absorption. It acts as a growth factor on certain tissues, through the activation of different signaling pathways increases expression of genes involved in cell cycle regulation such as CCND1, via JAK2-STAT3 pathway, or VEGFA, via MAPK1/3 and PI3K-AKT1 pathways. It may also play an apoptotic role via JAK2-STAT3 pathway and up-regulation of BIRC5 expression. It plays a pro-inflammatory role, in synergy with IL1B, by inducing NOS2 which promotes the production of IL6, IL8 and Prostaglandin E2, through a signaling pathway that involves JAK2, PI3K, MAP2K1/MEK1 and MAPK14/p38. In adaptive immunity, promotes the switch of memory T-cells towards T helper-1 cell immune responses. Increases CD4+CD25- T cells proliferation and reduces autophagy during TCR (T cell receptor) stimulation, through MTOR signaling pathway activation and BCL2 up-regulation.

This antiserum against Mouse Leptin has been tested for use in ELISA and immunoblotting.  Reactivity in other immunoassays is unknown.  Obesity associated leptin is the product of the OB gene and has been identified with Type II diabetes.This product has been assayed by immunoblot against tissue homogenates using HRP Goat-anti-Rabbit IgG [H&L] (code # 611-1302) and TMB as a substrate.  A working dilution range of 1:200 to 1:400 is suggested for this application. This product has been assayed by ELISA against recombinant mouse leptin using HRP Conjugated Anti-Rabbit IgG [H&L] (Goat) (code # 611-1302) and ABTS as a substrate for 30 minutes at room temperature.  A working dilution of 1:2,400 is suggested for this product. 

Type: Primary
Antigen: Leptin
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty