Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HIST1H3A Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
200-401-E81
76234-926EA 518 USD InStock
76234-926
Anti-HIST1H3A Rabbit Polyclonal Antibody
Antibodies
Histone H3 is one of the five main histone proteins involved in the structure of chromatin in eukaryotic cells. Histone proteins are highly post-translationally modified with Histone H3 being the most extensively modified of the five histones. The N-terminal tail of histone H3 protrudes from the globular nucleosome core and can undergo several different types of post-translational modification that influence cellular processes. Nucleosomes wrap and compact DNA into chromatin, limiting DNA accessibility to the cellular machineries which require DNA as a template. Histones thereby play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability. DNA accessibility is regulated via a complex set of post-translational modifications of histones, also called histone code, and nucleosome remodeling. Histone H3 Antibody is ideal for investigators involved in Cell Signaling, Epigenetics, Nuclear Signaling research and Signal Transduction research.

Histone H3 antibody has been tested for use in ELISA, IHC, and western blot. For western blots expect a band of approximately 15.4 kDa in size corresponding tothe Histone 3 protein. Specific conditions for reactivity should be optimized by the end user.

Type: Primary
Antigen: HIST1H3A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty