Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-VhH Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
200-401-GM6S
76234-932EA 150 USD InStock
76234-932
Anti-VhH Rabbit Polyclonal Antibody
Antibodies
Rabbit Anti-VhH antibody can be used to detect single domain VhH antibodies. The single-domain antibody (sdAb) is a small (12 - 14 kDa) antibody fragment that consists of a monomeric variable domain derived from the heavy chain, also called a VhH antibody. These heavy chain only Fab-like domains have activity that is similar to a whole antibody, and they are able to bind to a specific antigens. SdAb's are derived from camelid species that include llamas, alpacas and camels. Camelids produce both classical (containing heavy and light chain fragments) and non-classical antibody structures (containing only a heavy chain). VhH antibodies are the smallest functional antigen-binding fragment that occurs in nature and these are now being used in biotechnology as a novel antibody scaffold. The small size of the VhH single domain antibody makes it very attractive for use in diagnostic imaging and they have potential for therapeutic activity.

Anti-VhH antibody has been tested by ELISA and Western Blot. Specific conditions for reactivity should be optimized by the end user. Some cross-reactivity to E.coli proteins may be observed.

Type: Primary
Antigen: VhH
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty