Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IL1RA Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
209-401-303S
76234-946EA 148 USD InStock
76234-946
Anti-IL1RA Rabbit Polyclonal Antibody
Antibodies
Interleukin 1 receptor antagonist is secreted by various types of cells including immune, epithelial and adipocytes. IL 1Ra is used to treat rheumatoid arthritis. Anti-IL-1 RA antibody is ideal for investigators involved in Cancer and Immunology research.

This IgG fraction antibody of anti-Human IL-1 Receptor Antagonist has been tested for use in neutralizations, ELISA,  radioimmunoassays, western blotting, immunohistochemistry and flow cytometry.  Reactivity in other immunoassays is unknown.This product has been assayed by immunoblot using HRP Goat-anti-Rabbit IgG [H&L] (code # 611-1302) and TMB as a substrate.  A working dilution range of 1:400 to 1:800 is suggested for this application. This product has been assayed by ELISA against IL-1Receptor Antagonist using HRP Conjugated Anti-Rabbit IgG [H&L] (Goat) (code # 611-1302) and ABTS as a substrate for 30 minutes at room temperature.  A working dilution range of 1:500 to 1:1,000 is suggested for this product.  For use in ELISA formats, this antibody is  best used as the second antibody in combination with a monoclonal antibody as a capture antibody. This product has been assayed by immunohistochemistry.  A dilution of 1:400 or greater is suggested for this immunoassay.  Either paraffin fixation or cryofixation can be used for immunohistochemistry.  This product has been assayed by radioimmunoassay against antigen.  A dilution of 1:8,000 is suggested for this immunoassay.

Type: Primary
Antigen: IL1RA
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty