Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IL1R2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
209-401-305-0100
76234-948EA 518 USD InStock
76234-948
Anti-IL1R2 Rabbit Polyclonal Antibody
Antibodies
Interleukin 1 receptor type II binds to IL-1 alpha, IL-1 beta and IL1R1/IL1RA. Anti-IL-1 Receptor Type II antibody is ideal for investigators involved in Immunology and Stem Cell research.

This IgG fraction antibody of anti-Human IL-1 Receptor Type II (IL-1RII) has been tested for use in ELISA,  radioimmunoassays, flow cytometry and immunoprecipitation.  Reactivity in other immunoassays is unknown.   IL-1RII has been termed p68 or CD121 in some laboratories.  IL-1RII has been shown to bind IL-1, IL-1a and IL-1b.This product has been assayed for the ability to immunoprecipitate antigen.  A dilution range of 1:400 to 1:800 is suggested for this immunoassay.  For immunoprecipitation, pre-clearing with a non-specific rabbit IgG is helpful to reduce background.  This product has been assayed by immunoblot using HRP Goat-anti-Rabbit IgG [H&L] (code # 611-1302) and TMB as a substrate.  A working dilution range of 1:200 to 1:400 is suggested for this application. This product has been assayed by ELISA against soluble (extracellular) IL-1RII using HRP Conjugated Anti-Rabbit IgG [H&L] (Goat) (code # 611-1302) and ABTS as a substrate for 30 minutes at room temperature.  A working dilution range of 1:1,000 to 1:2000 is suggested for this product.  For use in ELISA formats, this antibody is best used as the second antibody in combination with a monoclonal antibody as a capture antibody.  This product has been assayed by radioimmunoassay against antigen.  A dilution of 1:8,000 is suggested for this immunoassay.  This product has been assayed for the ability to immunoprecipitate antigen.  A dilution range of 1:400 to 1:800 is suggested for this immunoassay.  For immunoprecipitation, pre-clearing with a non-specific rabbit IgG is helpful to reduce background.  This product has been assayed by immunoblot using HRP Goat-anti-Rabbit IgG [H&L] (code # 611-1302) and TMB as a substrate.  A working dilution range of 1:200 to 1:400 is suggested for this application. This product has been assayed by ELISA against soluble (extracellular) IL-1RII using HRP Conjugated Anti-Rabbit IgG [H&L] (Goat) (code # 611-1302) and ABTS as a substrate for 30 minutes at room temperature.  A working dilution range of 1:1,000 to 1:2000 is suggested for this product.  For use in ELISA formats, this antibody is best used as the second antibody in combination with a monoclonal antibody as a capture antibody.  This product has been assayed by radioimmunoassay against antigen.  A dilution of 1:8,000 is suggested for this immunoassay. 

Type: Primary
Antigen: IL1R2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty