Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TNFR-I Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
209-401-308S
76234-956EA 148 USD InStock
76234-956
Anti-TNFR-I Rabbit Polyclonal Antibody
Antibodies
Human TNF p55 Receptor Antibody recognizes TNFR1, the p55 soluble TNFR protein. sTNFRp55 is one of the two transmembrane receptors for tumor necrosis factor (TNF) with different molecular weight, 55 kD (p55) and 75 kD (p75), have been identified. The extracellular part of both receptors are shedded by proteolytic cleavage and exist as soluble receptors which bind to TNF in plasma and other biological fluids. sTNFR proteins have been implicated in certain leukemias. Anti-sTNFRp55 antibody is suitable for researchers in Cancer, Immunology, and Cell Biology.

Anti-TNF p55 Receptor (sTNFRp55) antibody has been tested for use in ELISA,  radioimmunoassays, immunoblotting, flow cytometry and immunoprecipitation.  Reactivity in other immunoassays is unknown. This product has been assayed for the ability to immunoprecipitate antigen and a dilution range of 1:400 to 1:800 is suggested.  For immunoprecipitation, pre-clearing with a non-specific rabbit IgG is helpful to reduce background.  This product has been assayed by immunoblot and a working dilution range of 1:200 to 1:400 is suggested for this application.  This product has been assayed by ELISA against soluble (extracellular) sTNFRp55 at a dilution range of 1:2,000 to 1:8,000.  For use in ELISA formats, this antibody is best used as the second antibody  in combination with a monoclonal antibody as a capture antibody.  This product has been assayed by radioimmunoassay against antigen.  A dilution of 1:8,000 is suggested for this immunoassay.

Type: Primary
Antigen: TNFR-I
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty