Would you like to log in again?
Anti-RFP Polyclonal Antibody
Supplier: Rockland Immunochemicals
200-101-379S 200-101-379
76234-958EA 559 USD InStock
76234-958 76234-960
Anti-RFP Polyclonal Antibody
Antibodies
Goat Anti-Red Fluorecent Protein Antibody
  • Purified IgG Fraction
  • No reaction was observed against Human, Mouse or Rat serum proteins
  • Western Blot
Antibodies to RFP (Discosoma spp.) are intended for use in immunological assays including ELISA, western blotting, immunofluorescence, and fluorescence activated cell sorting (FACS).
Polyclonal anti-RFP is designed to detect RFP and its variants. Optimal titers for applications should be determined by the researcher. Expect a band ~27kDa in Western Blot in the appropriate cell lysate or extract.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty