Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HbBs Mouse Monoclonal Antibody [Clone: 23E5.H6.G6.C1.H7.F7.G9.F6]

Supplier: Rockland Immunochemicals
200-301-GS5S
76234-984EA 148 USD InStock
76234-984
Anti-HbBs Mouse Monoclonal Antibody [Clone: 23E5.H6.G6.C1.H7.F7.G9.F6]
Antibodies
HbS antibodies detect the E6V mutant in the hemoglobin beta subunit. Functional adult hemoglobin (Hb) is a hetero tetramer composed of 2 alpha and 2 beta subunits (α2β2). Common isoform variants of hemoglobin include HbA, HbS, HbC, HbF, and HbA2. Hemoglobin S is the predominant hemoglobin in people with sickle cell disease. The alpha chain is normal. The disease-producing mutation exists in the beta chain, giving the molecule the structure, α2βS2. People who have one sickle mutant gene and one normal beta gene have sickle cell trait which is benign. Globin gene mutations affect the structure and expression levels of Hb. Sickle cell disease and the more benign sickle cell trait are observed in more than 100 million people globally. Perhaps the most significant mutation is the E6V in the beta subunit and the cause of SCD, but other relevant isoforms of Hb are observed. HbS antibody does not react to other forms of Hb. This antibody is ideal for investigators involved in Cardiovascular and developmental biology research.

Anti-Hemoglobin beta S (MOUSE) antibody is suitable for use in Western Blotting. Specific conditions of reactivity should be optimized by the end user. Expect a band of approximately 16 kDa in appropriate lysates.

Type: Primary
Antigen: HbBs
Clonality: Monoclonal
Clone: 23E5.H6.G6.C1.H7.F7.G9.F6
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG3 kappa
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty