Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HbG Mouse Monoclonal Antibody [Clone: 4B3.B5.F3.B7]

Supplier: Rockland Immunochemicals
200-301-GS6
76234-986EA 540 USD InStock
76234-986
Anti-HbG Mouse Monoclonal Antibody [Clone: 4B3.B5.F3.B7]
Antibodies
HbF antibodies detect the hemoglobin gamma isoform subunit. Functional alternate hemoglobin (Hb) is a hetero tetramer composed of 2 alpha and 2 gamma subunits (alpha-2 gamma-2). Hemoglobin F is elevated in newborns, reaching adult levels by 12 months. HbF levels are increased to as much as 5% to 10% in normal pregnancy. Sickle cell disease (SCD), thalassemias and hemoglobinopathies occur when aberrant forms of hemoglobin are expressed in children and adults. Hemoglobin variants arise from mutations in the globin genes and sickle cell disease and the more benign sickle cell trait are observed in more than 100 million people globally. HbF antibody does not react other forms of Hb. This antibody is ideal for investigators involved in Cardiovascular and developmental biology research.

Anti-Hemoglobin beta F (MOUSE) antibody is suitable for use in Western Blotting. Specific conditions of reactivity should be optimized by the end user. Expect a band of approximately 16 kDa.

Type: Primary
Antigen: HbG
Clonality: Monoclonal
Clone: 4B3.B5.F3.B7
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty