Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HbD Mouse Monoclonal Antibody [Clone: 21G1.F1.B9.G9.D11]

Supplier: Rockland Immunochemicals
200-301-GS8S
76234-996EA 148 USD InStock
76234-996
Anti-HbD Mouse Monoclonal Antibody [Clone: 21G1.F1.B9.G9.D11]
Antibodies
HbA-2 or hemoglobin delta subunit antibodies detect the delta-specific sequence in the hemoglobin delta-subunit found in HbA-2. Functional hemoglobin (Hb) is a hetero tetramer and the dominant form of Adult Hb is composed of 2 alpha and 2 beta subunits (α2β2). Hemoglobin A-2 (HbA-2) is a normal but minor variant of hemoglobin A that consists of two alpha and two delta chains (α2δ2). Hemoglobin A-2 may be increased in beta thalassemia or in people who are heterozygous for the beta thalassemia gene, and HbA2 is also linked to neurological disorders. HbA-2 form exists in small amounts in all adult humans (1.5-3.1% of all hemoglobin molecules) and is increased in people with Sickle-cell disease. Its normal biological role is not well understood. HbA-2 antibody does not react other forms of Hb including no cross-reaction to HbA or beta subunit. This antibody is ideal for investigators involved in Cardiovascular and developmental biology research.

Anti-Hemoglobin beta A-2 (MOUSE) antibody is suitable for use in Western Blotting. Specific conditions of reactivity should be optimized by the end user. Expect a band of approximately 16 kDa.

Type: Primary
Antigen: HbD
Clonality: Monoclonal
Clone: 21G1.F1.B9.G9.D11
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty