Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PAX6 Mouse Monoclonal Antibody [Clone: 7H10.D3.D4.G5]

Supplier: Rockland Immunochemicals
200-301-R22
76235-002EA 528 USD InStock
76235-002
Anti-PAX6 Mouse Monoclonal Antibody [Clone: 7H10.D3.D4.G5]
Antibodies
Paired box protein Pax-6 also known as aniridia type II protein (AN2) or oculorhombin is a transcription factor with key functions in the development of the eye, nose, central nervous system and pancreas. Pax6 further seems to be required for the differentiation of pancreatic islet alpha cells. PAX6 has been shown to bind to a promoter element in the glucagon, insulin and somatostatin promoters and regulates the specification of the ventral neuron subtypes by establishing the correct progenitor domains. Isoform 5a appears to function as a molecular switch that specifies target genes.
Anti-PAX6 antibody is ideal for researchers interested in developmental biology, stem cell research, cell growth and cancer research.

Anti-PAX6 antibody is suitable for ELISA and Immunofluorescence.  Specific conditions for reactivity should be optimized by the end user.  Expect a band approximately 46.7 kDa in size corresponding to PAX6 by western blotting in the appropriate cell lysate or extract.

Type: Primary
Antigen: PAX6
Clonality: Monoclonal
Clone: 7H10.D3.D4.G5
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty