Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MAP2K2 Mouse Monoclonal Antibody (FITC) [Clone: 19G10.F1.E2]

Supplier: Rockland Immunochemicals
200-302-GN8
76235-006EA 539 USD InStock
76235-006
Anti-MAP2K2 Mouse Monoclonal Antibody (FITC) [Clone: 19G10.F1.E2]
Antibodies
MEK2 antibodies detect the MEK2 isoform.  Mitogen-activated protein kinase kinase 2, also known as MEK2 or MKK2, is an integral component of the MAP kinase cascade that regulates cell growth and differentiation.   This pathway also plays a key role in synaptic plasticity in the brain.  Activated MEK 2 acts as a dual specificity kinase phosphorylating both a threonine and a tyrosine residue on MAP kinase.   MEK1 and MEK2 are about 80% identical to each other, and nearly identical within the kinase domain.   The MEK2 antibody is ideal for investigators involved in Neuroscience, Cell Signaling and Cancer Research.

Anti-MEK 2 FITC Conjugated (MOUSE) Antibody is suitable for use in Western Blotting and ELISA.  Specific conditions of reactivity should be optimized by the end user.  Expect a band of approximately 44 kDa.

Type: Primary
Antigen: MAP2K2
Clonality: Monoclonal
Clone: 19G10.F1.E2
Conjugation: FITC
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty