Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MAPK1 Mouse Monoclonal Antibody (DL680) [Clone: 3H9.A1.A8]

Supplier: Rockland Immunochemicals
200-344-GQ1
76235-048EA 539 USD InStock
76235-048
Anti-MAPK1 Mouse Monoclonal Antibody (DL680) [Clone: 3H9.A1.A8]
Antibodies
ERK2 antibodies detect the ERK2 isoform. Mitogen activated protein kinase 1, also known as MAPK1, ERK, or ERK2, is an integral component of the MAP kinase cascade that regulates cell growth and differentiation. ERK1 and ERK2 are activated by MEK1 and MEK2 in the B-raf signaling pathway resulting in its translocation to the nucleus where it phosphorylates nuclear targets. Human ERK1 and ERK2 are 84% identical in sequence and share common functionality in cells. Anti-ERK2 antibody is ideal for investigators involved in Neuroscience, Cell Signaling and Cancer Research.

Anti-ERK 2 (MOUSE) DyLight™680 conjugated antibody is suitable for use in ELISA and Western Blotting. Specific conditions of reactivity should be optimized by the end user. Expect a band of approximately 41 kDa.

Type: Primary
Antigen: MAPK1
Clonality: Monoclonal
Clone: 3H9.A1.A8
Conjugation: DL680
Epitope:
Host: Mouse
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty