Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MAP2K1 Mouse Monoclonal Antibody (DL800) [Clone: 13B6.G12]

Supplier: Rockland Immunochemicals
200-345-GQ0
76235-060EA 539 USD InStock
76235-060
Anti-MAP2K1 Mouse Monoclonal Antibody (DL800) [Clone: 13B6.G12]
Antibodies
MEK1 antibodies detect the MEK1 isoform.  Mitogen-activated protein kinase kinase 1, also known as MKK or MEK1, is an integral component of the MAP kinase cascade that regulates cell growth and differentiation.   This pathway also plays a key role in synaptic plasticity in the brain.  Activated MEK 1 acts as a dual specificity kinase phosphorylating both a threonine and a tyrosine residue on MAP kinase.   MEK1 and MEK2 are about 80% identical to each other, and nearly identical within the kinase domain.   The MEK1 antibody is ideal for investigators involved in Neuroscience, Cell Signaling and Cancer Research.

Anti-MEK 1 DyLight™800 Conjugated (MOUSE) Antibody is suitable for use in Western Blotting and ELISA.  Specific conditions of reactivity should be optimized by the end user.  Expect a band of approximately 43.5 kDa.

Type: Primary
Antigen: MAP2K1
Clonality: Monoclonal
Clone: 13B6.G12
Conjugation: DL800
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty