Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IL1A Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
209-401-302-0100
76235-118EA 518 USD InStock
76235-118
Anti-IL1A Rabbit Polyclonal Antibody
Antibodies
Interleukin 1 alpha is produced by activated macrophages. Anti-IL-1 alpha antibody is ideal for investigators involved in Immunology research.

This IgG fraction antibody of anti-Human IL-1a has been tested for use in neutralizations, ELISA,  immunohistochemistry, flow cytometry and immunoblotting.  It recognizes the 17,000 MW mature IL-1a.  Reactivity in other immunoassays is unknown.

Type: Primary
Antigen: IL1A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty