Would you like to log in again?
Ointment and Applicator Jars, Dukal
Supplier: Dukal
4237 4238
76235-134BX 322.36 USD InStock
76235-134 76235-132
Ointment and Applicator Jars, Dukal
Jars General Use Jars
These quality-crafted products are made from hospital grade stainless steel 18/8 (304) with seamless construction that can help to prevent bacteria growth and allows products to be cleaned with ease.
  • With handled lid
  • Autoclavable
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty